Assay Details
Target Gene Details
Entrez Gene ID: | 76251 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 6 like 2 |
Gene Aliases: |
0610007P08Rik, 1700019D06Rik, 9330134C04Rik, Rad26l, Sr278 |
Location: |
Chr.13:63813415-63900302 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ercc6l2 | NM_001013608.2 | NP_001013626.2 | ||
| NM_023507.3 | 14 | 2200 | NP_075996.2 | |
| AF217319.1 | 13 | 2017 | AAF73858.1 | |
| AK049874.1 | ||||
| BC032964.1 | 10 | 1228 | AAH32964.1 |

Genomic Map