Assay Details
Target Gene Details
Entrez Gene ID: | 218214 |
Gene Name: | lysine (K)-specific demethylase 1B |
Gene Aliases: |
4632428N09Rik, AI482520, Aof1 |
Location: |
Chr.13:47043471-47085279 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm1b | NM_172262.3 | NP_758466.1 | ||
| BC023917.1 | AAH23917.1 |
More Information
Set Membership: |
|

Genomic Map