Assay Details
Target Gene Details
Entrez Gene ID: | 108089 |
Gene Name: | ring finger protein 144A |
Gene Aliases: |
Rnf144, UIP4, Ubce7ip4 |
Location: |
Chr.12:26306794-26415321 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf144a | NM_001081977.2 | NP_001075446.1 | ||
| NM_080563.4 | NP_542130.1 | |||
| AF360998.1 | AAK51468.1 | |||
| AK038586.1 | BAC30058.1 | |||
| AK172904.1 | BAD32182.1 | |||
| BQ884505.1 | ||||
| BU055293.1 | ||||
| BY323271.1 | ||||
| BY753828.1 |
More Information
Set Membership: |
|

Genomic Map