Assay Details
Target Gene Details
Entrez Gene ID: | 625098 |
Gene Name: | solute carrier family 38, member 6 |
Gene Aliases: |
AW322671, EG625098 |
Location: |
Chr.12:73286794-73361998 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc38a6 | NM_001037717.3 | NP_001032806.2 | ||
| AK054341.1 | ||||
| AK140045.1 | BAE24222.1 | |||
| BC157917.1 | ||||
| BC157928.1 |
More Information
Set Membership: |
|

Genomic Map