Assay Details
Target Gene Details
Entrez Gene ID: | 20741 |
Gene Name: | spectrin beta, erythrocytic |
Gene Aliases: |
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219 |
Location: |
Chr.12:76580488-76710547 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptb | NM_013675.3 | 17 | 4330 | NP_038703.3 |
| AK147299.1 | 17 | 4331 | BAE27831.1 | |
| AK147684.1 | 17 | 3923 | BAE28073.1 | |
| AK147704.1 | 17 | 3923 | BAE28085.1 | |
| BC117711.2 | ||||
| BC129806.1 | 16 | 3663 | ||
| BC150783.1 | 16 | 3696 | ||
| M18641.1 | 2 | 477 | AAA40126.1 | |
| S66283.1 | 17 | 3806 | AAB28600.1 |

Genomic Map