Assay Details
Target Gene Details
Entrez Gene ID: | 68348 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 1F |
Gene Aliases: |
0610012A11Rik, AI327419, Serpina2, epserin |
Location: |
Chr.12:103688044-103695529 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina1f | NM_001164742.1 | 4 | 900 | NP_001158214.1 |
| NM_026687.2 | 5 | 1205 | NP_080963.2 | |
| AB233454.1 | 5 | 1205 | BAE45638.1 | |
| AK002574.1 | 4 | 1093 | BAB22199.1 | |
| AK165409.1 | 4 | 878 | BAE38167.1 | |
| BC119526.1 | 4 | 1033 |
More Information
Set Membership: |
|

Genomic Map