Assay Details
Target Gene Details
Entrez Gene ID: | 20741 |
Gene Name: | spectrin beta, erythrocytic |
Gene Aliases: |
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219 |
Location: |
Chr.12:76580488-76710547 on Build GRCm38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptb | NM_013675.3 | NP_038703.3 | ||
| AK147299.1 | BAE27831.1 | |||
| AK147684.1 | BAE28073.1 | |||
| AK147704.1 | BAE28085.1 | |||
| AK220551.1 | BAD90537.1 | |||
| BC117711.2 | ||||
| BC129806.1 | ||||
| BC150783.1 | ||||
| S66283.1 | 32 | 6531 | AAB28600.1 |
More Information
Set Membership: |
|

Genomic Map