Assay Details
Target Gene Details
Entrez Gene ID: | 74335 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 3 |
Gene Aliases: |
4432412E01Rik, AI182522, AW537713 |
Location: |
Chr.12:111803192-111813841 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrcc3 | NM_028875.2 | 5 | 611 | NP_083151.1 |
| AK014491.1 | 5 | 611 | BAB29391.1 | |
| BC043073.1 | 4 | 576 | AAH43073.1 |
More Information
Set Membership: |
|

Genomic Map