Assay Details
Target Gene Details
Entrez Gene ID: | 328059 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 15 |
Gene Aliases: |
Arpat |
Location: |
Chr.12:8528481-8599066 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a15 | NM_001038660.2 | 6 | 1458 | NP_001033749.1 |
| NM_177802.3 | 7 | 1015 | NP_808470.1 | |
| AK033481.1 | 7 | 1015 | BAC28311.1 | |
| AY795939.1 | AAX09981.1 |
More Information
Set Membership: |
|

Genomic Map