Assay Details
Target Gene Details
Entrez Gene ID: | 110893 |
Gene Name: | solute carrier family 8 (sodium/calcium exchanger), member 3 |
Gene Aliases: |
AW742262, Ncx3 |
Location: |
Chr.12:81197915-81333728 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc8a3 | NM_001167920.1 | NP_001161392.1 | ||
| NM_080440.3 | NP_536688.2 | |||
| AF453257.1 | AAL39160.1 | |||
| AK139260.1 | BAE23935.1 | |||
| AK165197.1 | BAE38073.1 | |||
| BC052435.1 | AAH52435.1 | |||
| BC080862.1 | AAH80862.1 |
More Information
Set Membership: |
|

Genomic Map