Assay Details
Target Gene Details
Entrez Gene ID: | 69123 |
Gene Name: | enoyl-Coenzyme A delta isomerase 3 |
Gene Aliases: |
1810022C23Rik, Kseg2 |
Location: |
Chr.13:34946614-34963884 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eci3 | NM_026947.4 | NP_081223.1 | ||
| AK007578.1 | ||||
| AK131590.1 | BAE20705.1 | |||
| BC014724.1 | AAH14724.1 | |||
| FJ687155.1 |
More Information
Set Membership: |
|

Genomic Map