Assay Details
Target Gene Details
Entrez Gene ID: | 66761 |
Gene Name: | RIKEN cDNA 4933417A18 gene |
Gene Aliases: |
- |
Location: |
Chr.13:34924405-34956218 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933417A18Rik | NM_025750.3 | 5 | 967 | NP_080026.2 |
| AK016778.1 | 5 | 932 | BAB30424.1 | |
| AK016838.1 | 5 | 969 | BAB30456.1 | |
| BC100450.1 | AAI00451.1 |
Target Gene Details
Entrez Gene ID: | 69123 |
Gene Name: | enoyl-Coenzyme A delta isomerase 3 |
Gene Aliases: |
1810022C23Rik, Kseg2 |
Location: |
Chr.13:34946614-34963884 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eci3 | NM_026947.4 | NP_081223.1 | ||
| AK007578.1 | ||||
| AK131590.1 | BAE20705.1 | |||
| BC014724.1 | AAH14724.1 | |||
| FJ687155.1 |
More Information
Set Membership: |
|

Genomic Map