Assay Details
Target Gene Details
Entrez Gene ID: | 104886 |
Gene Name: | RAB15, member RAS oncogene family |
Gene Aliases: |
2310012G06Rik, AI840042 |
Location: |
Chr.12:76797960-76822761 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab15 | NM_134050.4 | NP_598811.3 | ||
| AB232610.1 | BAF02872.1 | |||
| AK158637.1 | BAE34591.1 | |||
| AK158768.1 | BAE34652.1 | |||
| BC013790.1 | AAH13790.1 | |||
| BC027769.1 | AAH27769.1 |
More Information
Set Membership: |
|

Genomic Map