Assay Details
Target Gene Details
Entrez Gene ID: | 58185 |
Gene Name: | radical S-adenosyl methionine domain containing 2 |
Gene Aliases: |
2510004L01Rik, Vig1, cig5 |
Location: |
Chr.12:26442743-26456478 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
More Information
Set Membership: |
|

Genomic Map