Assay Details
Target Gene Details
Entrez Gene ID: | 217734 |
Gene Name: | protein-O-mannosyltransferase 2 |
Gene Aliases: |
A830009D15Rik, AW046274 |
Location: |
Chr.12:87106861-87147902 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pomt2 | NM_153415.3 | 18 | 2324 | NP_700464.2 |
| AF246235.1 | 18 | 1928 | AAL85627.1 | |
| AK043573.1 | 18 | 2025 | ||
| AK043713.1 | 18 | 1866 | ||
| AK050915.1 | 18 | 1893 | BAC34458.1 | |
| AK076813.1 | 18 | 2327 | ||
| AY090481.1 | 18 | 2324 | AAM12047.1 | |
| AY090482.1 | 18 | 1924 | AAM12048.1 | |
| BC052045.1 | 17 | 1977 | AAH52045.1 | |
| BC062965.1 | 17 | 1977 | AAH62965.1 |
More Information
Set Membership: |
|

Genomic Map