Assay Details
Target Gene Details
Entrez Gene ID: | 16195 |
Gene Name: | interleukin 6 signal transducer |
Gene Aliases: |
5133400A03Rik, AA389424, BB405851, CD130, D13Ertd699e, gp130 |
Location: |
Chr.13:112464070-112506860 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il6st | NM_010560.3 | NP_034690.3 | ||
| AK142637.1 | BAE25142.1 | |||
| BC058679.1 | AAH58679.1 | |||
| M83336.1 | AAA37723.1 | |||
| X62646.1 | CAA44515.1 |
More Information
Set Membership: |
|

Genomic Map