Assay Details
Target Gene Details
Entrez Gene ID: | 16553 |
Gene Name: | kinesin family member 13A |
Gene Aliases: |
4930505I07Rik, mKIAA4109 |
Location: |
Chr.13:46749087-46931300 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif13a | NM_010617.2 | 30 | 3820 | NP_034747.2 |
| AB037923.2 | 30 | 3821 | BAB16346.1 | |
| AK029721.1 | 12 | 1456 | BAC26583.1 | |
| AK220490.1 | 7 | 774 | BAD90288.1 |
More Information
Set Membership: |
|

Genomic Map