Assay Details
Target Gene Details
Entrez Gene ID: | 75398 |
Gene Name: | mitochondrial ribosomal protein L32 |
Gene Aliases: |
0610033O15Rik |
Location: |
Chr.13:14610301-14613037 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl32 | NM_029271.2 | NP_083547.1 | ||
| AB049650.1 | BAB40855.1 | |||
| AF279255.1 | AAK69482.1 | |||
| AK002753.1 | BAB22330.1 | |||
| AK031791.1 | 1 | 112 | ||
| AK078324.1 | 1 | 110 | ||
| BC052214.1 | AAH52214.1 |
More Information
Set Membership: |
|

Genomic Map