Assay Details
Target Gene Details
Entrez Gene ID: | 13039 |
Gene Name: | cathepsin L |
Gene Aliases: |
1190035F06Rik, Ctsl1, MEP, fs, nkt |
Location: |
Chr.13:64363214-64370883 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsl | NM_009984.3 | 4 | 414 | NP_034114.1 |
| AF121837.1 | 4 | 378 | AAD32136.1 | |
| AF121838.1 | 4 | 378 | AAD32137.1 | |
| AF121839.1 | 4 | 378 | AAD32138.1 | |
| AK002221.1 | 4 | 414 | BAB21945.1 | |
| AK011589.1 | 4 | 414 | BAB27719.1 | |
| AK049466.1 | 4 | 415 | BAC33761.1 | |
| AK136341.1 | 3 | 255 | BAE22939.1 | |
| AK145051.1 | 4 | 415 | BAE26206.1 | |
| AK147142.1 | 4 | 416 | BAE27712.1 | |
| AK150452.1 | 4 | 415 | BAE29572.1 | |
| AK151169.1 | 4 | 415 | BAE30172.1 | |
| AK153076.1 | 4 | 415 | BAE31701.1 | |
| AK153417.1 | 4 | 415 | BAE31977.1 | |
| AK159511.1 | 4 | 415 | BAE35143.1 | |
| AK160100.1 | 4 | 415 | BAE35627.1 | |
| AK161538.1 | 4 | 415 | BAE36450.1 | |
| AK165397.1 | 4 | 415 | BAE38161.1 | |
| AK169394.1 | 4 | 415 | BAE41141.1 | |
| AK169712.1 | 4 | 415 | BAE41323.1 | |
| AK190042.1 | ||||
| AK190644.1 | ||||
| AK199491.1 | ||||
| BC068163.1 | 4 | 400 | AAH68163.1 | |
| J02583.1 | 4 | 391 | AAA37445.1 | |
| X04392.1 | 1 | 68 | CAA27980.1 | |
| X06086.1 | 4 | 400 | CAA29470.1 |
More Information
Set Membership: |
|

Genomic Map