Assay Details
Target Gene Details
Entrez Gene ID: | 64139 |
Gene Name: | cathepsin M |
Gene Aliases: |
1600027J17Rik, AI324100, Catm |
Location: |
Chr.13:61535743-61541839 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsm | NM_022326.3 | NP_071721.2 | ||
| AF202528.1 | 2 | 162 | AAF68224.1 | |
| AF456463.1 | AAO27847.1 | |||
| AK005428.1 | BAB24022.1 | |||
| AK005550.1 | BAB24116.1 | |||
| AY014777.1 | AAK00506.1 |
More Information
Set Membership: |
|

Genomic Map