Assay Details
Target Gene Details
Entrez Gene ID: | 218268 |
Gene Name: | eukaryotic translation initiation factor 4E family member 1B |
Gene Aliases: |
Eif4eloo, Gm273 |
Location: |
Chr.13:54783927-54788459 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4e1b | NM_001033269.3 | NP_001028441.1 | ||
| NM_001039683.2 | NP_001034772.1 | |||
| NM_001286178.1 | NP_001273107.1 | |||
| NM_001286179.1 | NP_001273108.1 | |||
| NM_001286180.1 | NP_001273109.1 | |||
| NR_104406.1 | ||||
| NR_104407.1 | ||||
| AK139577.1 | BAE24071.1 | |||
| BC139143.1 | ||||
| BC139144.1 | ||||
| DQ279473.1 | ABB88894.1 | |||
| DQ279474.1 | ABB88895.1 | |||
| DQ279475.1 | ABB88896.1 | |||
| DQ279476.1 | ABB88897.1 | |||
| DQ279477.1 | ABB88898.1 | |||
| DQ279478.1 | ABB88899.1 | |||
| DQ279479.1 | ABB88900.1 | |||
| DV653822.1 | ||||
| DV659735.1 | ||||
| DV662251.1 | ||||
| DV662389.1 |
More Information
Set Membership: |
|

Genomic Map