Assay Details
Target Gene Details
Entrez Gene ID: | 218506 |
Gene Name: | mitochondrial ribosomal protein S27 |
Gene Aliases: |
2610028H14Rik |
Location: |
Chr.13:99344767-99415562 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps27 | NM_173757.3 | NP_776118.2 | ||
| AK076008.1 | BAC36116.1 | |||
| AK087686.1 | ||||
| BC049149.1 | AAH49149.1 |
More Information
Set Membership: |
|

Genomic Map