Assay Details
Target Gene Details
Entrez Gene ID: | 56835 |
Gene Name: | cathepsin R |
Gene Aliases: |
AI428350 |
Location: |
Chr.13:61159215-61164188 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsr | NM_020284.1 | NP_064680.1 | ||
| AF245399.1 | AAF90051.1 | |||
| AK005429.1 | BAB24023.1 | |||
| AK014432.1 | BAB29345.1 | |||
| AY014778.1 | AAK00507.1 | |||
| AY943668.1 | AAX39778.1 | |||
| BC100338.1 | AAI00339.1 |
More Information
Set Membership: |
|

Genomic Map