Assay Details
Target Gene Details
Entrez Gene ID: | 218629 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 29 |
Gene Aliases: |
3732415M03, AU043276, Ddxx, E130202M19Rik |
Location: |
Chr.13:112927530-112969187 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhx29 | NM_172594.2 | 7 | 832 | NP_766182.2 |
| AK042497.1 | BAC31274.2 | |||
| AK049530.1 | 7 | 905 | BAC33796.1 | |
| AK087450.1 | 7 | 1146 | BAC39880.1 | |
| BC057112.1 | 7 | 905 | AAH57112.1 | |
| BC082319.1 | 7 | 895 | AAH82319.1 |
More Information
Set Membership: |
|

Genomic Map