Assay Details
Target Gene Details
Entrez Gene ID: | 57266 |
Gene Name: | chemokine (C-X-C motif) ligand 14 |
Gene Aliases: |
1110031L23Rik, 1200006I23Rik, AI414372, BMAC, BRAK, KS1, Kec, MIP-2g, MIP2gamma, NJAC, Scyb14, bolekine |
Location: |
Chr.13:56288643-56296792 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxcl14 | NM_019568.2 | NP_062514.2 | ||
| AB041614.1 | BAA95097.1 | |||
| AF144754.1 | AAF66694.1 | |||
| AF152377.1 | AAD34157.1 | |||
| AF192557.1 | AAF03753.1 | |||
| AF252873.1 | AAF65570.1 | |||
| AF352784.1 | AAK52902.1 | |||
| AF352785.1 | AAK52903.1 | |||
| AK004615.1 | BAB23411.1 | |||
| AK014351.1 | BAB29292.1 | |||
| AK076112.1 | BAC36192.1 | |||
| AK155356.1 | BAE33213.1 | |||
| AU035952.1 | ||||
| BC079661.1 | AAH79661.1 | |||
| BC147274.1 | ||||
| BC147275.1 |
More Information
Set Membership: |
|

Genomic Map