Assay Details
Target Gene Details
Entrez Gene ID: | 320806 |
Gene Name: | G elongation factor, mitochondrial 2 |
Gene Aliases: |
6530419G12Rik, A930009M04Rik, EF-G2mt, EFG2, MST027, RRF2mt |
Location: |
Chr.13:97137937-97181196 on Build GRCm38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfm2 | NM_001146043.2 | NP_001139515.1 | ||
| AK044750.1 | 1 | 816 | ||
| AK052633.1 | 5 | 2110 | ||
| AK161797.1 | BAE36579.1 |
Target Gene Details
Entrez Gene ID: | 15212 |
Gene Name: | hexosaminidase B |
Gene Aliases: |
- |
Location: |
Chr.13:97176331-97198357 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hexb | NM_010422.2 | NP_034552.1 | ||
| AK151149.1 | BAE30155.1 | |||
| AK159134.1 | BAE34846.1 | |||
| AK159178.1 | BAE34877.1 | |||
| AK160560.1 | BAE35874.1 | |||
| AK168825.1 | BAE40652.1 | |||
| CD775639.1 | ||||
| U07633.1 | AAA18776.1 | |||
| Y00964.1 | CAA68781.1 |
More Information
Set Membership: |
|

Genomic Map