Assay Details
Target Gene Details
Entrez Gene ID: | 320806 |
Gene Name: | G elongation factor, mitochondrial 2 |
Gene Aliases: |
6530419G12Rik, A930009M04Rik, EF-G2mt, EFG2, MST027, RRF2mt |
Location: |
Chr.13:97137937-97181196 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfm2 | NM_001146043.2 | 13 | 1250 | NP_001139515.1 |
| NM_001271463.1 | 13 | 1250 | NP_001258392.1 | |
| NM_001271464.1 | 12 | 1169 | NP_001258393.1 | |
| NM_001271465.1 | 13 | 1277 | NP_001258394.1 | |
| NM_177266.5 | 13 | 1283 | NP_796240.3 | |
| AK028450.1 | 13 | 1236 | BAC25958.1 | |
| AK035819.1 | 13 | 1189 | ||
| AK044371.1 | 12 | 1170 | BAC31889.1 | |
| AK083441.1 | 13 | 1218 | ||
| AK161797.1 | 13 | 1186 | BAE36579.1 | |
| AK165871.1 | 13 | 1244 | BAE38428.1 | |
| BC027341.1 | 13 | 1234 | AAH27341.1 | |
| BC138233.1 | 13 | 1181 | ||
| BC145208.1 | 13 | 1181 |
More Information
Set Membership: |
|

Genomic Map