Assay Details
Target Gene Details
Entrez Gene ID: | 15212 |
Gene Name: | hexosaminidase B |
Gene Aliases: |
- |
Location: |
Chr.13:97176331-97198357 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hexb | NM_010422.2 | 8 | 1068 | NP_034552.1 |
| AK151149.1 | 8 | 1005 | BAE30155.1 | |
| AK159134.1 | 8 | 1021 | BAE34846.1 | |
| AK159178.1 | 8 | 1042 | BAE34877.1 | |
| AK160560.1 | 8 | 1037 | BAE35874.1 | |
| AK168825.1 | 8 | 1042 | BAE40652.1 | |
| AK194154.1 | ||||
| AK212361.1 | ||||
| U07633.1 | 8 | 1040 | AAA18776.1 | |
| Y00964.1 | 8 | 981 | CAA68781.1 |
More Information
Set Membership: |
|

Genomic Map