Assay Details
Target Gene Details
Entrez Gene ID: | 70930 |
Gene Name: | nucleolar protein 8 |
Gene Aliases: |
4921532D18Rik, 5730412B09Rik |
Location: |
Chr.13:49653078-49679016 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol8 | NM_001271397.1 | NP_001258326.1 | ||
| NR_073167.1 | ||||
| NR_073168.1 | ||||
| AK017551.1 | ||||
| AK147172.1 | BAE27736.1 | |||
| BC094941.1 | AAH94941.1 | |||
| BC125575.1 | ||||
| BC125579.1 |
More Information
Set Membership: |
|

Genomic Map