Assay Details
Target Gene Details
Entrez Gene ID: | 105364 |
Gene Name: | expressed sequence AW495222 |
Gene Aliases: |
- |
Location: |
Chr.13:93706100-93719175 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AW495222 | NR_045086.1 | 2 | 439 | |
| AK046172.1 | 2 | 440 | ||
| BB314344.2 | 2 | 262 | ||
| BB644732.1 | 2 | 437 | ||
| BE656108.1 | 2 | 322 |
Target Gene Details
Entrez Gene ID: | 74129 |
Gene Name: | dimethylglycine dehydrogenase precursor |
Gene Aliases: |
1200014D15Rik, AI787269 |
Location: |
Chr.13:93674436-93752823 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmgdh | NM_028772.3 | 12 | 1873 | NP_083048.1 |
| AK004755.1 | 12 | 1874 | BAB23536.1 | |
| BC024126.1 | 8 | 1156 | AAH24126.1 | |
| BC089599.1 | 12 | 1876 | AAH89599.1 |
More Information
Set Membership: |
|

Genomic Map