Assay Details
Target Gene Details
Entrez Gene ID: | 76856 |
Gene Name: | cation channel, sperm associated 3 |
Gene Aliases: |
4921522D01Rik, CACRC |
Location: |
Chr.13:55784579-55824942 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Catsper3 | NM_001252487.1 | NP_001239416.1 | ||
| NM_001252488.1 | NP_001239417.1 | |||
| NM_029772.4 | NP_084048.1 | |||
| AK014942.1 | BAB29631.1 | |||
| AY263400.1 | AAP21831.1 | |||
| HM179540.1 | ||||
| HM179541.1 |
More Information
Set Membership: |
|

Genomic Map