Assay Details
Target Gene Details
Entrez Gene ID: | 26401 |
Gene Name: | mitogen-activated protein kinase kinase kinase 1 |
Gene Aliases: |
MAPKKK1, MEKK1, Mekk |
Location: |
Chr.13:111746433-111818269 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map3k1 | NM_011945.2 | NP_036075.2 | ||
| AB014614.1 | BAA85878.1 | |||
| AF117340.1 | AAD25049.1 |
More Information
Set Membership: |
|

Genomic Map