Assay Details
Target Gene Details
Entrez Gene ID: | 218461 |
Gene Name: | phosphodiesterase 8B |
Gene Aliases: |
B230331L10Rik, C030047E14Rik |
Location: |
Chr.13:95024105-95250397 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde8b | NM_001170669.1 | NP_001164140.1 | ||
| NM_172263.2 | NP_758467.2 | |||
| AK036090.1 | BAC29302.1 | |||
| AK043680.1 | ||||
| AK045991.1 | ||||
| AK083166.1 | BAC38791.1 | |||
| AK138301.1 | BAE23616.1 | |||
| BC032286.1 | AAH32286.1 | |||
| EF644173.1 |
More Information
Set Membership: |
|

Genomic Map