Assay Details
Target Gene Details
Entrez Gene ID: | 66671 |
Gene Name: | cyclin H |
Gene Aliases: |
6330408H09Rik, AI661354, AV102684, AW538719 |
Location: |
Chr.13:85189377-85213726 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnh | NM_023243.5 | NP_075732.1 | ||
| AF287135.1 | AAF90198.1 | |||
| AK007910.1 | BAB25343.1 | |||
| AK078059.1 | BAC37112.1 | |||
| AK137849.1 | BAE23508.1 | |||
| BC038861.1 | AAH38861.1 |
More Information
Set Membership: |
|

Genomic Map