Assay Details
Target Gene Details
Entrez Gene ID: | 59079 |
Gene Name: | Erbb2 interacting protein |
Gene Aliases: |
1700028E05Rik, Erbin, mKIAA1225 |
Location: |
Chr.13:103818786-103920586 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erbb2ip | NM_001005868.2 | 24 | 4422 | NP_001005868.1 |
| NM_001289473.1 | 23 | 4278 | NP_001276402.1 | |
| NM_001289474.1 | NP_001276403.1 | |||
| NM_001289475.1 | 23 | 4305 | NP_001276404.1 | |
| AF263743.1 | 3 | 1159 | AAF77047.1 | |
| AK029054.1 | 13 | 3104 | BAC26267.1 | |
| AK051733.1 | 3 | 937 | BAC34742.1 | |
| AK122473.1 | 23 | 3879 | BAC65755.1 | |
| BC005691.1 | 3 | 403 | ||
| BC028256.1 | 4 | 1865 | AAH28256.1 | |
| BC141178.1 | ||||
| BC141188.1 | 24 | 4131 | ||
| BC145498.1 | 23 | 4014 | ||
| BC145509.1 | 23 | 3987 | ||
| EU233994.1 | 26 | 5372 |
More Information
Set Membership: |
|

Genomic Map