Assay Details
Target Gene Details
Entrez Gene ID: | 26385 |
Gene Name: | G protein-coupled receptor kinase 6 |
Gene Aliases: |
Gprk6 |
Location: |
Chr.13:55445072-55460927 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grk6 | NM_001038018.4 | 3 | 382 | NP_001033107.1 |
| NM_001112711.2 | 3 | 382 | NP_001106182.1 | |
| NM_001286063.1 | 3 | 362 | NP_001272992.1 | |
| NM_001286064.1 | 3 | 382 | NP_001272993.1 | |
| NM_001286065.1 | 3 | 382 | NP_001272994.1 | |
| NM_001286066.1 | 3 | 382 | NP_001272995.1 | |
| NM_011938.4 | 3 | 382 | NP_036068.2 | |
| AF040747.1 | 3 | 252 | AAC09268.1 | |
| AF040748.1 | 3 | 252 | AAC09269.1 | |
| AF040749.1 | 3 | 252 | AAC09270.1 | |
| AK051405.1 | 3 | 363 | ||
| AK146119.1 | 3 | 257 | BAE26914.1 | |
| AK161142.1 | BAE36212.1 | |||
| BC075719.1 | 3 | 263 | AAH75719.1 | |
| CJ179645.1 | ||||
| Y15797.1 | 3 | 204 | CAA75788.1 | |
| Y15798.1 | 3 | 226 | CAA75789.1 | |
| Y15799.1 | 3 | 209 | CAA75790.1 | |
| Y15800.1 | 3 | 204 | CAA75791.1 |
More Information
Set Membership: |
|

Genomic Map