Assay Details
Target Gene Details
Entrez Gene ID: | 28114 |
Gene Name: | NOL1/NOP2/Sun domain family member 2 |
Gene Aliases: |
D13Wsu123e, Misu |
Location: |
Chr.13:69612016-69635780 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsun2 | NM_145354.5 | NP_663329.3 | ||
| AI787627.1 | ||||
| AK030124.1 | 1 | 61 | BAC26795.1 | |
| AK150631.1 | BAE29720.1 | |||
| AK151917.1 | BAE30795.1 | |||
| AK154789.1 | ||||
| AK171399.1 | ||||
| AK188384.1 | ||||
| AK197440.1 | ||||
| AK215502.1 | ||||
| BC025549.1 | ||||
| CJ092700.1 | ||||
| DQ490066.1 |
More Information
Set Membership: |
|

Genomic Map