Assay Details
Target Gene Details
Entrez Gene ID: | 28114 |
Gene Name: | NOL1/NOP2/Sun domain family member 2 |
Gene Aliases: |
D13Wsu123e, Misu |
Location: |
Chr.13:69612016-69635780 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsun2 | NM_145354.5 | 13 | 1535 | NP_663329.3 |
| AK030124.1 | 12 | 1420 | BAC26795.1 | |
| AK075999.1 | 3 | 314 | BAC36110.1 | |
| AK150631.1 | 13 | 1489 | BAE29720.1 | |
| AK151917.1 | 13 | 1489 | BAE30795.1 | |
| AK154789.1 | 13 | 1527 | ||
| AK171399.1 | 13 | 1492 | ||
| AK205395.1 | 3 | 248 | ||
| AK206159.1 | 1 | 147 | ||
| BC013625.1 | 9 | 1007 | AAH13625.1 | |
| BC025549.1 | ||||
| DQ490066.1 | 13 | 1498 |
More Information
Set Membership: |
|

Genomic Map