Assay Details
Target Gene Details
Entrez Gene ID: | 72935 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 41 |
Gene Aliases: |
2900024F02Rik, AA958953, ABS, AI324246 |
Location: |
Chr.13:55530410-55536658 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx41 | NM_134059.2 | 11 | 1323 | NP_598820.2 |
| AK151427.1 | 11 | 1197 | BAE30391.1 | |
| AK156957.1 | 11 | 1191 | BAE33914.1 | |
| AK168043.1 | 11 | 1196 | BAE40025.1 | |
| BC011308.1 | 11 | 1177 | AAH11308.1 | |
| BE864921.1 | 2 | 113 | ||
| CX231996.1 | 4 | 516 |

Genomic Map