Assay Details
Target Gene Details
Entrez Gene ID: | 59079 |
Gene Name: | Erbb2 interacting protein |
Gene Aliases: |
1700028E05Rik, Erbin, mKIAA1225 |
Location: |
Chr.13:103818786-103920586 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erbb2ip | NM_001005868.2 | 3 | 490 | NP_001005868.1 |
| NM_001289473.1 | 3 | 490 | NP_001276402.1 | |
| NM_001289474.1 | 3 | 490 | NP_001276403.1 | |
| NM_001289475.1 | 3 | 490 | NP_001276404.1 | |
| AK122473.1 | 3 | 169 | BAC65755.1 | |
| AK169529.1 | 3 | 419 | ||
| BC141178.1 | 3 | 199 | ||
| BC141188.1 | 3 | 199 | ||
| BC145498.1 | 3 | 199 | ||
| BC145509.1 | 3 | 199 | ||
| BM933013.1 | 1 | 108 | ||
| BY745732.1 | 3 | 419 | ||
| EU233994.1 | 5 | 1246 |
More Information
Set Membership: |
|

Genomic Map