Assay Details
Target Gene Details
Entrez Gene ID: | 59079 |
Gene Name: | Erbb2 interacting protein |
Gene Aliases: |
1700028E05Rik, Erbin, mKIAA1225 |
Location: |
Chr.13:103818786-103920586 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erbb2ip | NM_001005868.2 | NP_001005868.1 | ||
| NM_001289473.1 | NP_001276402.1 | |||
| NM_001289474.1 | NP_001276403.1 | |||
| NM_001289475.1 | NP_001276404.1 | |||
| AK122473.1 | BAC65755.1 | |||
| AK169529.1 | ||||
| BB376299.2 | ||||
| BC141178.1 | ||||
| BC141188.1 | ||||
| BC145498.1 | ||||
| BC145509.1 | ||||
| EU233994.1 |
More Information
Set Membership: |
|

Genomic Map