Assay Details
Target Gene Details
Entrez Gene ID: | 23894 |
Gene Name: | general transcription factor II H, polypeptide 2 |
Gene Aliases: |
44kDa, BTF2 p44, BTF2-p44, Btf2p44, p44 |
Location: |
Chr.13:100459786-100492610 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf2h2 | NM_022011.4 | NP_071294.3 | ||
| AB221674.1 | ||||
| AK010543.1 | ||||
| AK078297.1 | BAC37209.1 | |||
| AK145720.1 | ||||
| AK154739.1 | ||||
| AK158705.1 | ||||
| BC016231.1 | AAH16231.1 | |||
| BC053382.1 | AAH53382.1 | |||
| BY277671.1 | ||||
| DV662169.1 |
More Information
Set Membership: |
|

Genomic Map