Assay Details
Target Gene Details
Entrez Gene ID: | 23986 |
Gene Name: | enoyl-Coenzyme A delta isomerase 2 |
Gene Aliases: |
Acbd2, Drs1, Hca88, Peci |
Location: |
Chr.13:34977748-35027089 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eci2 | NM_001110331.1 | NP_001103801.1 | ||
| NM_011868.3 | NP_035998.2 | |||
| NR_073427.1 | ||||
| AF153613.1 | AAD34174.1 | |||
| AK009478.1 | BAB26315.2 | |||
| AK170778.1 | BAE42023.1 | |||
| AK218002.1 | ||||
| BC001983.1 | AAH01983.1 | |||
| BQ176032.1 |
More Information
Set Membership: |
|

Genomic Map