Assay Details
Target Gene Details
Entrez Gene ID: | 26914 |
Gene Name: | H2A histone family, member Y |
Gene Aliases: |
H2AF12M, mH2a1, macroH2A1 |
Location: |
Chr.13:56073622-56135626 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2afy | NM_001159513.1 | NP_001152985.1 | ||
| NM_001159514.1 | NP_001152986.1 | |||
| NM_001159515.1 | NP_001152987.1 | |||
| NM_012015.2 | NP_036145.1 | |||
| AB071988.2 | BAB68541.1 | |||
| AF171080.1 | AAD53745.1 | |||
| AK039274.1 | BAC30302.1 | |||
| AK143498.1 | ||||
| BB633156.1 | ||||
| BC006955.1 | AAH06955.1 |
More Information
Set Membership: |
|

Genomic Map