Assay Details
Target Gene Details
Entrez Gene ID: | 17260 |
Gene Name: | myocyte enhancer factor 2C |
Gene Aliases: |
5430401D19Rik, 9930028G15Rik, AV011172, Mef2 |
Location: |
Chr.13:83504034-83667079 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mef2c | NM_001170537.1 | 1 | 289 | NP_001164008.1 |
| NM_025282.3 | 1 | 289 | NP_079558.1 | |
| AK009139.1 | 1 | 129 | BAB26099.1 | |
| AK132573.1 | 1 | 118 | BAE21236.1 | |
| CJ155464.1 | 1 | 289 |

Genomic Map