Assay Details
Target Gene Details
Entrez Gene ID: | 13206 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 4 |
Gene Aliases: |
AV206478, Mvh, VASA |
Location: |
Chr.13:112598333-112652629 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx4 | NM_001145885.1 | 17 | 1429 | NP_001139357.1 |
| NM_010029.2 | 16 | 1351 | NP_034159.1 | |
| AK014844.1 | 16 | 1342 | BAB29578.1 | |
| AK133366.1 | 17 | 1422 | BAE21618.1 | |
| BC137601.1 | 16 | 1405 | ||
| BC144760.1 | 16 | 1405 | ||
| D14859.1 | 13 | 1124 | BAA03584.1 | |
| JQ923486.1 | 16 | 1379 |

Genomic Map