Assay Details
Target Gene Details
Entrez Gene ID: | 13206 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 4 |
Gene Aliases: |
AV206478, Mvh, VASA |
Location: |
Chr.13:112598333-112652629 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx4 | NM_001145885.1 | NP_001139357.1 | ||
| NM_010029.2 | NP_034159.1 | |||
| AK014844.1 | BAB29578.1 | |||
| AK133366.1 | BAE21618.1 | |||
| BC137601.1 | ||||
| BC144760.1 | ||||
| BE860399.1 | ||||
| JQ923486.1 |

Genomic Map