Assay Details
Target Gene Details
Entrez Gene ID: | 14756 |
Gene Name: | glycosylphosphatidylinositol specific phospholipase D1 |
Gene Aliases: |
6330541J12Rik, AW546131 |
Location: |
Chr.13:24943152-24991936 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpld1 | NM_008156.2 | 14 | 1438 | NP_032182.2 |
| AF050666.1 | 14 | 1383 | AAC77799.1 | |
| AK004957.2 | 13 | 1142 | BAB23698.1 | |
| AY081194.1 | 13 | 1165 | AAL87452.1 | |
| AY333960.1 | 13 | 1122 | AAP93132.1 | |
| BC019146.1 | 13 | 1151 | AAH19146.1 |
More Information
Set Membership: |
|

Genomic Map