Assay Details
Target Gene Details
Entrez Gene ID: | 17260 |
Gene Name: | myocyte enhancer factor 2C |
Gene Aliases: |
5430401D19Rik, 9930028G15Rik, AV011172, Mef2 |
Location: |
Chr.13:83504034-83667079 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mef2c | NM_001170537.1 | NP_001164008.1 | ||
| NM_025282.3 | NP_079558.1 | |||
| AK009139.1 | BAB26099.1 | |||
| AK077603.1 | ||||
| AK132573.1 | BAE21236.1 | |||
| BC026841.1 | AAH26841.1 | |||
| BC037731.1 | AAH37731.1 | |||
| BC057650.1 | AAH57650.1 | |||
| L13171.1 |
More Information
Set Membership: |
|

Genomic Map