Assay Details
Target Gene Details
Entrez Gene ID: | 20706 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 9b |
Gene Aliases: |
1600019A21Rik, R86, SPI-CI, Spi10, ovalbumin |
Location: |
Chr.13:33027414-33040559 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb9b | NM_011452.2 | NP_035582.1 | ||
| AK005491.1 | BAB24076.1 | |||
| U96705.2 | AAB57817.2 |
More Information
Set Membership: |
|

Genomic Map